Search Thermo Fisher Scientific
Catalog Number | Length (English) Shaft | Length (Metric) Shaft | Tip Type |
---|---|---|---|
FIS23-600-968 | 3.15 in. | 80 mm | Contoured Pediatric |
FIS23-600-950 | 3.15 in. | 80 mm | Minitip flocked |
FIS23-600-951 | 3.15 in. | 80 mm | Regular flocked |
FIS23-600-955 | 3.94 in. | 100 mm | Ultra minitip/pediatric |
FIS23-600-957 | 3.94 in. | 100 mm | Regular flocked |
FIS23-600-952 | 3.94 in. | 100 mm | Flexible Minitip |