Hansel™ Stains
Actual product may vary
Lide Labs
Hansel™ Stains
For the examination of the nasal secretions to differentiate an allergic condition from a bacterial infection
Have Questions?
Catalog NumberQuantity
FIS23-233-801500 mL
Catalog number FIS23-233-801
Price (SGD)
-
Quantity:
500 mL
  • One-minute stain
  • For secretion cytology
  • Stain eosinophils red; neutrophils, epithelial (exfoliated cells) and bacteria blue; and mast cells or basophiles red and blue
  • Can be adapted for staining various secretions: urinary, conjunctival, salivary, gastrointestinal, nasal, sinus, aural and bronchial
  • Determine elevated levels of eosinophils in urine (eosinophiluria) which can aid in diagnosis of many associated clinical conditions
  • Cannot be used for blood staining
Specifications
For Use With (Application)Secretion cytology stains eosinophils red, neutrophils, epithelial (exfoliated cells) and bacteria blue, and mast cells or basophiles red and blue
Quantity500 mL
Product TypeHansel Stain
Unit SizeEach