Specifications
Quantity250 mL/Bottle
Includes250mL gram crystal violet , 250mL stabilized iodine , 250mL decolorizer and 250mL safranin
FormatBottle
For Use With (Application)Laboratory chemicals
ColorReddish Violet
Stain TypeGram Stain
Product TypeGram Stain Kit
Unit SizeEach