Check Order Status
Conductivity Standard, 200,000 μS/cm (200 mS/cm) at 25°C, Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 200,000 μS/cm (200 mS/cm) at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS224920-32
1 L
Catalog number
FIS224920-32
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity Standard
Name Note
200 mS/cm (200, 000 μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
83.0
View more