Check Order Status
Conductivity Standard, 150,000 μS/cm (150 mS/cm) at 25°C, Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 150,000 μS/cm (150 mS/cm) at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS224915-32
1 L
Catalog number
FIS224915-32
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity Standard
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
89.0
CAS Max %
93.0
View more