Check Order Status
Conductivity Standard, 200 μS/cm at 25°C (95 ppm TDS as NaCl), Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 200 μS/cm at 25°C (95 ppm TDS as NaCl), Ricca Chemical
Have Questions?
Change view
Quantity:
1 L
500 mL
Catalog number
FIS2238-32
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity/TDS Standard
Packaging
Natural Poly Bottle
CAS
7647-14-5
Concentration or Composition Notes
95ppm as NaCl
CAS
7732-18-5
View more