Histoplast Paraffin Wax
Actual product may vary
Histoplast Paraffin Wax
Three blends to accommodate all specimen types
Have Questions?
Catalog NumberProduct TypeMelting Point
FIS22-900-700Paraffin, Histoplast PE54° to 57°C
Catalog number FIS22-900-700
Price (SGD)
-
Product Type:
Paraffin, Histoplast PE
Melting Point:
54° to 57°C
When purity matters, only the finest will do.
  • Formulated using raw materials of only the highest purity
  • Is not sticky or brittle while sectioning
  • Provides consistent tissue ribbons every time
Histoplast PE
For general use and all histology applications
Histoplast IM
For use on specimens requiring more thorough infiltration (large and dense specimens)
Histoplast LP
A lower melting-point formula for improved ribboning with less compression
  • Chemical Stability: Stable under normal temperatures and pressures
  • Conditions to Avoid: Excess heat
  • Incompatibilities with Other Materials: Strong oxidizing agents
  • Hazardous Decomposition Products: Product does not decompose at ambient temperatures
  • Hazardous Polymerization: Will not occur
Specifications
Melting Point54° to 57°C
Product TypeParaffin, Histoplast PE
Quantity1kg
For Use With (Application)General use and all histology applications
Unit SizeCase