Paraffin Wax Scrapers: Replacement Blades
Actual product may vary
Dynamic Diagnostics
Paraffin Wax Scrapers: Replacement Blades
Replacement blades for Dynamic Diagnostics Safety, Mini, Floor and Maxi scrapers
Have Questions?
Catalog NumberLength (English)For Use With (Equipment)
FIS22-899-2631.5 in.Safety and mini scraper
Catalog number FIS22-899-263
Price (SGD)
137.00
Pack
Length (English):
1.5 in.
For Use With (Equipment):
Safety and mini scraper
Price (SGD)
137.00
Pack
For scrapers, see cat. no. 22-899-262.
Specifications
Product TypeReplacement Blade for Scrapers
For Use With (Equipment)Safety and mini scraper
Length (English)1.5 in.
Length (Metric)3.81 cm
MaterialSteel
Unit SizePack