Graduated Medicine Cups
Actual product may vary
Medline
Graduated Medicine Cups
Translucent cups suitable for dispensing both liquid and dry medications
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-666-47030 mL
Catalog number FIS22-666-470
Price (SGD)
260.00
Case
Capacity (Metric):
30 mL
Price (SGD)
260.00
Case
  • Disposable
  • Plastic
  • 1 oz.
  • Graduated from 1–8 drams, 0.125–1 fluid ounce, 0.5 teaspoon–2 tablespoons and 2.5–30mL and cc
  • 100 per sleeve
Specifications
SterilityNon-sterile
Capacity (Metric)30 mL
ColorTranslucent
Product TypeMedicine Cup
Capacity (English)1.01 oz.
MaterialPlastic
DisposableSingle-use
For Use With (Application)Dispensing both liquid and dry medications
Latex FreeYes
Capacity30 mL
Unit SizeCase