Manual Differential Counters
Actual product may vary
LW Scientific
Manual Differential Counters
Designed for blood cell counting
Have Questions?
Catalog NumberDimensions (L x W x H)Weight (Metric)
FIS22-500-10027.9 x 10.1 x 7.62cm1.36kg
Catalog number FIS22-500-100
Price (SGD)
652.00
Each
Dimensions (L x W x H):
27.9 x 10.1 x 7.62cm
Weight (Metric):
1.36kg
Price (SGD)
652.00
Each
  • Durable and acid-resistant
  • High-impact plastic body
  • Right and left reset knobs
  • Non-slip bottom
  • Strip chart of mature and immature forms
  • Bell alarm for 100th counted cell
  • Five and Eight key counters available
Specifications
No. of KeysFive key
Dimensions (L x W x H)27.9 x 10.1 x 7.62cm
Weight (Metric)1.36kg
For Use With (Application)Blood cell counting
Unit SizeEach