Search Thermo Fisher Scientific
| Catalog Number | Volume | Product Type | Quantity |
|---|---|---|---|
| FIS22-363-602 | 0.1 μL / 10 μL | Inoculating Loop and Needle, Flexible | 5000/Cs. |
| FIS22-363-605 | 10 μL | Inoculating Loop and Needle, Rigid | 1 Ea. |
| FIS22-363-604 | 1 μL | Inoculating Loop and Needle, Rigid | 9600/Cs. |
| FIS22-363-606 | 10 μL | Inoculating Loop and Needle, Rigid | 40 Pk. per Cs. |
| FIS22-363-607 | 10 μL | Inoculating Loop and Needle, Rigid | 9600/Cs. |
| FIS22-363-603 | 1 μL | Inoculating Loop and Needle, Rigid | 250/Cs. |
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.