Latex-Free Cohesive Flexible Bandages
Latex-Free Cohesive Flexible Bandages
Actual product may vary
Latex-Free Cohesive Flexible Bandages
Free of natural rubber proteins and allergens.
Have Questions?
Catalog NumberColorDimensions (L x W)
FIS22-363-135Blue5 yd. x 3 in.
Catalog number FIS22-363-135
Price (SGD)
-
Color:
Blue
Dimensions (L x W):
5 yd. x 3 in.
  • Maintain consistent compression without slipping or restricting mobility
  • High porosity; water-resistant
  • Lightweight, porous material allows skin to breathe for ultimate comfort
  • Adhere to themselves — no clips or fasteners needed
  • Available in a wide range of colors and widths
  • Length: 5 yd. (4.6m)
Specifications
MaterialNon-woven Fabric
Product TypeCohesive Flexible Bandage
Length (English)5 yd.
Length (Metric)4.6 m
Width (Metric)7.6 cm
Width (English)3 in.
Dimensions (L x W)5 yd. x 3 in.
WaterproofYes
For Use With (Application)Prevent muscle sprain and injury, bind up after hemostasia by compression, without slipping or restricting mobility
ColorBlue
AdhesiveSelf Adhesive
Latex FreeYes
Certifications/ComplianceCE
Unit SizeCS