SUN-SRi™ Luer-Slip Syringes
Actual product may vary
Thermo Scientific
SUN-SRi™ Luer-Slip Syringes
Siliconized and assembled with scale and tip shield.
Have Questions?
Catalog NumberVolume (Metric)Product TypeSterility
FIS22-3143832 mLLuer-slip SyringeNon-sterile
Catalog number FIS22-314383
Price (SGD)
740.00
Case
Volume (Metric):
2 mL
Product Type:
Luer-slip Syringe
Sterility:
Non-sterile
Price (SGD)
740.00
Case
Syringe, Chemically resistant, inert polypropylene and polyethylene, Luer Slip, Bulk
  • Chemically resistant, inert polypropylene and polyethylene
  • No rubber plunger seals or silicone lubricants that may cause sample contamination
  • Bulk packaged, choice or sterile or nonsterile
  • LUER-Slip connection
Specifications
MaterialPlastic
Product TypeLuer-slip Syringe
DisposableSingle-use
Volume (Metric)2 mL
Needle Point StyleCentered
Connection TypeLeur Slip