Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Racks
▸
Microtube Racks
▸
80 Well Micro Tube Rack, Lavender
Actual product may vary
Bio Plas
80 Well Micro Tube Rack, Lavender
Designed with 80 wells in a 16 x 5 configuration with each well numbered
Have Questions?
Catalog Number
Positions
FIS22-313630
80
Catalog number
FIS22-313630
Price (SGD)
58.00
Pack
Add to cart
Positions:
80
Price (SGD)
58.00
Pack
Add to cart
For microcentrifuge tubes
Surface allows convenient sample identification
L x D: 9 x 2.06 in.
Well Diameter: 10.5mm
Specifications
Color
Lavender
Holds
Microtubes (0.5, 1.2, and 2 mL)
Positions
80
Length (Metric)
22.8 cm
Length (English)
9 in.