▸
Environmental Samplers
▸
General Purpose Sample Bags
▸
Reclosable White Specimen Bags
Actual product may vary
Minigrip
Reclosable White Specimen Bags
Reclosable zipper lock bags
Have Questions?
Catalog Number
Length (English)
Width (English)
FIS22-310-032
6 in.
4 in.
Catalog number
FIS22-310-032
Price (SGD)
77.00
Case
Add to cart
Length (English):
6 in.
Width (English):
4 in.
Price (SGD)
77.00
Case
Add to cart
White block design with writable area
2 mil thickness
Specifications
Closure Type
Zip Seal
Thickness (English)
2 mil
Length (English)
6 in.
Width (English)
4 in.