Emesis Containment Bag
Actual product may vary
Minigrip
Emesis Containment Bag
Special Emesis bag featuring two outside hand flaps and a liquid-tight safety seal
Have Questions?
Catalog NumberDimensions (L x W)
FIS22-310-01110 x 10 in.
Catalog number FIS22-310-011
Price (SGD)
944.00
Case
Dimensions (L x W):
10 x 10 in.
Price (SGD)
944.00
Case
Perfect for bedside applications at surgery and oncology centers or emergency rooms and for odd-sized or specialty transport needs.
  • Extra-wide opening is designed for ease-of-use
  • Hand flaps on each side of the bag help to protect against splatter
  • Adhesive strip seals liquid tight, ready for safe disposal
  • Biohazard logo and illustrated instructions printed on the bag
  • A special score provides easy access for later analysis of content
  • Bag may be written on to identify contents
  • 100% disposable
  • Used by hospital personnel, patients, EMTs, paramedics, ambulance personnel, and nursing home staff
Specifications
DisposableSingle-use
For Use With (Application)Hospital personnel, patients, EMT's paramedics, ambulance personnel and nursing home staff
IncludesLiquid tight adhesive closure, score opening (for lab testing), biohazard symbol
Dimensions (L x W)10 x 10 in.
Width (English)10 in.
Height (English)10 in.
Height (Metric)25.4 cm
Product TypeEmesis Containment Bag
Width (Metric)25.4 cm
Unit SizeCase