Test Strips
Actual product may vary
Test Strips
Test Strips
LaMotte
Test Strips
For chlorine, iodine and QAC testing
Have Questions?
Catalog NumberTypeQuantity
FIS22-307-864QAC Test Strip100 Pk.
Catalog number FIS22-307-864
Price (SGD)
-
Type:
QAC Test Strip
Quantity:
100 Pk.
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm
  • Chemically treated paper strips produce color changes in as little as 5 seconds
  • Simply dip strip into sample
  • Compare strip color to color chart located on plastic vial
  • Plastic, waterproof vials are labeled with test instructions
  • Desiccant cap
Test drinking water, industrial water for boiling and cooling, laundry /sanitation water and wastewater
Specifications
For Use With (Application)Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing
IncludesColor chart, plastic waterproof opaque vial, desiccant cap
Quantity100 Pk.
Range50-100-200-400 ppm
Shelf Life1 Year
TypeQAC Test Strip
Unit SizePack