▸
Environmental Samplers
▸
General Purpose Sample Bags
▸
Red Line™ Plain Reclosable Zipper Bags
Actual product may vary
Minigrip
Red Line™ Plain Reclosable Zipper Bags
Leak-resistant bags with or without hang holes
Have Questions?
Catalog Number
Dimensions (L x W)
Thickness (English)
FIS22-306-400
8 x 10 in. (20.3 x 25.4cm)
4 mil
Catalog number
FIS22-306-400
Price (SGD)
472.00
Case
Add to cart
Dimensions (L x W):
8 x 10 in. (20.3 x 25.4cm)
Thickness (English):
4 mil
Price (SGD)
472.00
Case
Add to cart
Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
With reinforced zipper and side seams
Thickness of 4 mil
With or without hang holes
Clear
Packaged in DispenserBag™
Specifications
Closure Type
Zipper
Color
Clear
Thickness (English)
4 mil
Dimensions (L x W)
8 x 10 in. (20.3 x 25.4cm)