Reloc ZIPPIT™ Plain Reclosable Zipper Bags
Actual product may vary
Minigrip
Reloc ZIPPIT™ Plain Reclosable Zipper Bags
Have Questions?
Catalog NumberDimensions (L x W)
FIS22-306-3326 x 6 in.
Catalog number FIS22-306-332
Price (SGD)
182.00
Case
Dimensions (L x W):
6 x 6 in.
Price (SGD)
182.00
Case
  • Economical
  • Color: Clear
  • Pack Quantity: 100
  • Inner Case Quantity: 1000
  • Inner Case Weight: 14.2 lb.
Specifications
MaterialPlastic
Thickness (English)4 mil
Dimensions (L x W)6 x 6 in.
Product TypeZip-seal Storage Bag
Unit SizeCase