Check Order Status
Kimble™ Glass Racks for Westergren Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Glass Racks for Westergren Tubes
Reduces the chance of bio hazardous contamination
Have Questions?
Catalog Number
Positions
For Use With (Application)
FIS22-295694
10
Westergren and Wintrobe Tubes
Catalog number
FIS22-295694
Price (SGD)
-
Positions:
10
For Use With (Application):
Westergren and Wintrobe Tubes
Racks feature leveling bubbles and adjustment screws
Tubes are held firmly in a perpendicular position to ensure accurate readings.
Specifications
Autoclavable
Not Autoclavable
Material
Glass
Positions
10
For Use With (Application)
Westergren and Wintrobe Tubes