Cover Glasses: Circles
Cover Glasses: Circles
Actual product may vary
Fisherbrand
Cover Glasses: Circles
Made of finest optical borosilicate glass, with uniform thickness and size
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Diameter (Metric)
FIS22-2932320.17 to 0.25 mm12 mm
FIS12-545-81#1.5 (0.16 to 0.19 mm)12 mm
FIS12-546-10.17 to 0.25 mm22 mm
FIS12-546-20.17 to 0.25 mm25 mm
FIS12-545-101#1 (0.13 to 0.16 mm)22 mm
FIS12-545-102#1 (0.13 to 0.16 mm)25 mm
Catalog number FIS22-293232
Price (SGD)
-
Thickness (Metric):
0.17 to 0.25 mm
Diameter (Metric):
12 mm
Made of finest optical borosilicate glass, with uniform thickness and size
Circular shape.
  • Corrosion-resistant
  • Lint-free and ready to use
  • Available in squares, circles and rectangles
Specifications
Product TypeCoverslip
ShapeCircle
MaterialBorosilicate Glass
Thickness (Metric)0.17 to 0.25 mm
Diameter (Metric)12 mm
For Use With (Equipment)Microscopes
Unit SizePK