Wide Mouth Color-Code Safety Labeled Wash Bottles
Actual product may vary
Bel-Art
Wide Mouth Color-Code Safety Labeled Wash Bottles
Easy-to-read labeling printed bottle promoting safe working environment
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)For Use With (Application)
FIS22-288641500 mLEthanol
FIS22-273739500 mLMethyl Ethyl Ketone
FIS14-512-38500 mLSodium Hypochlorite
Catalog number FIS22-288641
Price (SGD)
245.00
Case
Capacity (Metric):
500 mL
For Use With (Application):
Ethanol
Price (SGD)
245.00
Case
Easy-to-read labeling printed bottle promoting safe working environment
  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Made from solvent-resistant low-density polyethylene.
  • Polypropylene closure with leakproof design.
  • Pictogram instructions promote universal safety regardless of setting.
  • Use LYOB only with solvents compatible with LDPE.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorClear
For Use With (Application)Ethanol
MaterialLDPE
SterilityNon-sterile
Certifications/ComplianceOSHA 29 CFR 1910.1200
Closure IncludedYes
DisposableReusable
GraduatedNo
MouthWide
ShapeCylindrical
Product TypeWash Bottle
Chemical LabelEthanol
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypePolypropylene Closure
Through the Closure DispensingYes