Circular Pipet Stand
Actual product may vary
Bel-Art
Circular Pipet Stand
For safe and orderly storage of up to 28 pipets in a space-saving stand
Have Questions?
Catalog NumberPositions
FIS22-27550328
Catalog number FIS22-275503
Price (SGD)
94.00
Each
Positions:
28
Price (SGD)
94.00
Each
For safe and orderly storage of up to 28 pipets in a space-saving stand
  • Polypropylene stand has a two-section top plate
  • Holes in the lower plate are 4mm in diameter and support the tips of the pipets as they are held in the stand
Specifications
AutoclavableAutoclavable
ColorWhite
HoldsPipets
MaterialPolypropylene
Positions28
ShapeRound
StackableNo
SterilityNon-sterile
Width (English)7 in.
Height (English)7 in.
Product TypePipet Rack
ShelvesNone
Height (Metric)17.78 cm
Width (Metric)17.8 cm
Unit SizeEach