Ubbelohde Calibrated UBC Series Viscometers
Actual product may vary
Cannon
Ubbelohde Calibrated UBC Series Viscometers
Suspended level instrument for measurement of kinematic viscosity.
Have Questions?
Catalog NumberViscosity RangeApproximate ConstantASTM NumbersSize
FIS22-274003200 to 1000 cSt1 cSt/sec.33
Catalog number FIS22-274003
Price (SGD)
867.00
Each
Viscosity Range:
200 to 1000 cSt
Approximate Constant:
1 cSt/sec.
ASTM Numbers:
3
Size:
3
Price (SGD)
867.00
Each
  • Glass capillary with expanded uncertainty
  • Minimum sample volume of 0.37 oz.(11mL)
  • Provided with certificate of calibration and instruction sheet
  • Measure transparent Newtonian liquids
Specifications
Product TypeUbbelohde Viscometer
Viscosity Range200 to 1000 cSt
CalibratedYes
Approximate Constant1 cSt/sec.
ASTM Numbers3
Certifications/ComplianceASTM D445, ASTM D446, ISO 3104, ISO 3105, calibration certificate
IncludesInstruction sheet, calibration certificate
MaterialBorosilicate Glass
Depth (English) Bath9.5 in.
Depth (Metric) Bath24.1 cm
Precision±0.2%
For Use With (Application)For measurement of kinematic viscosity of transparent Newtonian liquids by suspended level principle
Size3
Volume (Metric)11 mL
Unit SizeEach