CryoBox™ Freezer Racks
Actual product may vary
CryoBox™ Freezer Racks
CryoBox™ Freezer Racks
CryoBox™ Freezer Racks
Thermo Scientific
CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsDesignDimensions (L x W x H)
FIS22-2730327 CryoBoxes/RackVertical14.3 x 14 x 70.5 cm
5036-00044 CryoBoxes/RackVertical14.3. x 14 x 22.5 cm
Catalog number FIS22-273032
Price (SGD)
2,274.00
Case
Holds:
7 CryoBoxes/Rack
Design:
Vertical
Dimensions (L x W x H):
14.3 x 14 x 70.5 cm
Price (SGD)
2,274.00
Case
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
  • Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
  • Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
  • Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Specifications
ColorSilver
Dimensions (L x W x H)14.3 x 14 x 70.5 cm
Holds7 CryoBoxes/Rack
DesignVertical
MaterialStainless Steel
TypeFreezer Rack
Unit SizeCase