Spinbar™ Magnetic Retriever
Actual product may vary
Spinbar™ Magnetic Retriever
Spinbar™ Magnetic Retriever
Bel-Art
Spinbar™ Magnetic Retriever
Reaches down drains for lost bars
Have Questions?
Catalog NumberLength (Metric)
FIS22-26974645.7 cm
Catalog number FIS22-269746
Price (SGD)
50.00
Each
Length (Metric):
45.7 cm
Price (SGD)
50.00
Each
Reaches down drains for lost bars
  • 0.38 in. (9.5 mm) dia. polyethylene pick-up rod
  • With powerful Alnico magnet sealed in one end
  • Handy hang-up grip on other end
  • Offers convenient stir bar retrieval for bars up to 1 in. long
Specifications
Dimensions (L x W x H)9.5 mm x 45.7 cm
For Use With (Application)Offers convenient stir bar retrieval for bars up to 1 in. long
For Use With (Equipment)Stirring bars
IncludesHigh-strength alnico sealed-in magnet, hang-up loop on one end, 0.95 cm polyethylene pick-up rod
Length (English)18 in.
Diameter (Metric)9.5 mm
Length (Metric)45.7 cm
MaterialPolyethylene
Quantity1
ShapeCylinder
TypeSpinbar™ Magnetic Retriever
Unit SizeEach