Evaporating Dish
Evaporating Dish
Actual product may vary
Evaporating Dish
Evaporating Dish
Evaporating Dish
Bel-Art
Evaporating Dish
Manufactured from polypropylene. Evaporating dish is smooth with easy pour spout for use in a variety of laboratory tasks.
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-261902175 mL
Catalog number FIS22-261902
Price (SGD)
146.00
PK
Add to cart
Capacity (Metric):
175 mL
Price (SGD)
146.00
PK
Add to cart
  • Useful for titration and evaporation
  • Resists acids and alkalis
  • Autoclavable at 121°C (250°F)
  • Can also be used as a reusable weighing dish
  • 100mm diameter x 42mm deep, holds up to 175mL
Specifications
AutoclavableAutoclavable
Bottom ShapeFlat
Capacity (Metric)175 mL
ColorNatural
MaterialPolypropylene
ShapeRound
SterilityNon-sterile
Surface TreatmentNon-treated
TypeEvaporating Dish
Volume (Metric) Working175 mL
For Use With (Application)Titration and Evaporation
Interior FinishSmooth
Height (Metric)4.2 cm
Diameter (English)4 in.
Diameter (Metric)10 cm
Unit SizePK