TFE Forceps
TFE Forceps
Actual product may vary
Bel-Art
TFE Forceps
Rugged forceps are chemical and corrosion resistant and withstand autoclaving at 199°C (390°F)
Have Questions?
Catalog number FIS22-261826
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Optimized for handling higher temperature small items or balance weights
  • Forceps tips are square, 1.6mm (1/16 in.) wide, with flat surfaces
  • 4.5 in. (114mm) long
Specifications
Tip StyleSquare
Length (English)4.48 in.
MaterialFluorocarbon
Length (Metric)114 mm
Product TypeForceps
Quantity (No. per Case)12/Case
TypePlastic Forceps
StyleStandard
Unit SizeEA