Check Order Status
Bellows Samplers
Actual product may vary
Bel-Art
Bellows Samplers
Provide quick and easy sampling with one-handed operation
Have Questions?
Catalog Number
Volume (Metric)
Length (Metric)
FIS22-261785
50 mL
50 cm
Catalog number
FIS22-261785
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
50 mL
Length (Metric):
50 cm
Provide quick and easy sampling with one-handed operation
Long tapered tip with 4 mm opening
FDA grade polyethylene construction
Graduated to indicate volume collected
Specifications
Material
Polyethylene
Sterility
Non-sterile
Graduated
Yes
Quantity
5/Pk.
Tip Style
Tapered
Packaging Type
Bag
Color
Translucent
Volume (Metric)
50 mL
Length (Metric)
50 cm
Type
Bellows Sampler
Disposable
Single-use