Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Blood, Hematology and Coagulation Testing Products
▸
Blood Specimen Collection
▸
Capillary Blood Collection Tubes
▸
Color-Coded Capillary Tubes
Actual product may vary
Fisherbrand
Color-Coded Capillary Tubes
Excellent for use in microhematocrit determinations
Have Questions?
Change view
Catalog Number
Product Type
Closure Color
FIS22-260950
Heparinized Tube
Red
FIS22-260943
Non-heparinized Tube
Blue
2 Options
Catalog number
FIS22-260950
Price (SGD)
9.00
Pack
Add to cart
Product Type:
Heparinized Tube
Closure Color:
Red
Price (SGD)
9.00
Pack
Add to cart
Excellent for use in microhematocrit determinations
Ends can be sealed by flame, sealing clay or caps
Manufactured from low-taper, soft glass
Tubes are packed in plastic vials with reclosable snap-lock caps to maintain cleanliness
Red band on tube indicates heparinization; blue band indicates nonheparinized tube
Interior surface of heparinized tubes is evenly coated with ammonium heparin (4 to 7 USP units), which will not interfere with sodium determination
Specifications
Color
Red
Product Type
Heparinized Tube
Additive
Ammonium Heparin
Material
Soda Lime Glass
Sterility
Non-sterile
Closure Type
Tube Sealant
Capacity (Metric)
70 μL
Diameter (Metric) Inner
1.1 to 1.2 mm
Closure Color
Red
Length (Metric)
75 mm