Specifications
Capacity1 oz., 35 mL
Capacity (Metric)29.6 mL
Capacity (English)1 oz.
ColorWhite
Diameter (English)1.69 in.
For Use With (Application)Storage of Liquid or Dry Stock, for use with Samples
IncludesScrew Cap
MaterialPolypropylene
SterilityNon-sterile
Closure ColorWhite
Closure IncludedYes
DisposableReusable
GraduatedNo
MouthWide, Threaded
ShapeRound
Height (Metric)37.3 mm
Product TypeStorage Jar
Height (English)1.64 in.
Chemical LabelUnlabeled
Closure LinerPolyethylene Foam
Closure MaterialPolypropylene
Diameter (Metric)43 mm
Closure TypeScrew Cap