Broken Glass Disposal Box
Actual product may vary
Bel-Art
Broken Glass Disposal Box
Ruggedly constructed, puncture-resistant glass disposal receptacles safely collect broken and discarded laboratory glass. Boxes are clearly labeled, and highly visible.
Have Questions?
Catalog NumberCapacity (English)
FIS22-260-0992.3 gal.
Catalog number FIS22-260-099
Price (SGD)
277.00
Pack
Capacity (English):
2.3 gal.
Price (SGD)
277.00
Pack
Ruggedly constructed, puncture-resistant glass disposal receptacles safely collect broken and discarded laboratory glass. Boxes are clearly labeled, and highly visible.
  • Sturdy corrugated cardboard receptacle is supplied with a 2 mil thick polyethylene bag to contain broken glass fragments and spilled liquids
  • Printed with eye-catching graphics in four languages (English, German, French, Spanish)
  • Packaged flat to save space and shipping costs and is quickly assembled
  • Does not qualify as a DOT container
  • Safe puncture resistant receptacles for broken glassware
  • Floor model holds 40 lb. (18 kg) of waste
Specifications
Capacity (Metric)10 L
Capacity (English)2.3 gal.
ColorGreen
Dimensions (L x W x H)12 x 12 x 27 in. (30 x 30 x 69 cm)
Height (Metric)69 cm
Length (Metric)30 cm
Width (Metric)30 cm
AutoclavableNot Autoclavable
Width (English)12 in.
Length (English)12 in.
Height (English)27 in.
MaterialCorrugated Cardboard
Product TypeSafety Container
Unit SizePack