Check Order Status
Disposable Type II Soda Lime Glass Pasteur Pipets in Stand-Up Case
Actual product may vary
Disposable Type II Soda Lime Glass Pasteur Pipets in Stand-Up Case
Tip openings range from 0.03 to 0.06 in. (0.80 to 1.40mm)
Have Questions?
Change view
Catalog Number
Length (Metric)
FIS22-230490
22.86 cm
FIS22-230-482
14.6 cm
2 Options
Catalog number
FIS22-230490
Price (SGD)
-
Length (Metric):
22.86 cm
The tip openings measure from 0.03 to 0.06 in. (0.80 to 1.40mm).
High-quality flint glass pipets in convenient stand-up case
Saves bench space
Approximate volume: 2mL
Can be cotton plugged (plugs not included)
Specifications
Certifications/Compliance
ASTM E732
Material
Flint Glass
Includes
Stand-up case
Length (English)
9 in.
Volume (English)
0.067 oz.
Volume (Metric)
2 mL
Length (Metric)
22.86 cm
Type
Pasteur Pipet
Disposable
Single-use