KV3000 and KV4000 Constant-Temperature Viscosity Baths
KV3000 and KV4000 Constant-Temperature Viscosity Baths
Actual product may vary
KV3000 and KV4000 Constant-Temperature Viscosity Baths
KV3000 and KV4000 Constant-Temperature Viscosity Baths
Koehler Instrument
KV3000 and KV4000 Constant-Temperature Viscosity Baths
Simple push-button operation
Have Questions?
Catalog NumberModel
FIS22-172-590KV3000
Catalog number FIS22-172-590
Price (SGD)
-
Model:
KV3000
Kinematic viscosity is of primary importance in the design and selection of a wide range of petroleum products.
  • Constant temperature bath series with advanced temperature control circuitry and integrated timing features for convenient, accurate glass capillary viscometry determinations
  • Reliable temperature, simple push-button controls, and accommodation of up to seven capillary viscometers
  • Connection of the built-in cooling coil to tap water or a recirculating water chiller facilitates temperature control at ambient or below ambient temperatures
Integrated Timing
  • Model KV3000: Seven individual start/stop timers with displays Model KV4000: Integrated LCD microcomputer with start/stop buttons and retention of viscometer tube constants, automatic calculation and display in viscosity units or seconds; temperature control between ambient and 150°C (302°F), and down to 10°C (50°F); 12 or 18 in. bath depth
Specifications:
  • Electrical Reqts: 115V 50/60Hz, single phase 12.6A
  • Shipping Weight: 101 lb. (45.9kg) Dimensions: 29L x 25W x 34 in.H (73.7 x 63.5 x 86.4cm)
Specifications
Controller TypeMicroprocessor
Certifications/ComplianceASTM D445, D2170, D6074, D6158, IP 71, 319, ISO 3104, DIN 51550, FTM 791-305, NF T 60-100
Temperature RangeAmbient to 150°C
ModelKV3000
Dimensions (L x W x H)29 x 25 x 34 in. (73.7 x 63.5 x 86.4 cm)
Length (English)29 in.
Length (Metric)73.7 cm
Width (English)25 in.
Width (Metric)63.5 cm
Height (English)34 in.
Height (Metric)86.4 cm
Depth (English)12 in.
Depth (Metric)30.5 cm
Shipping Weight (English)101 lb.
Shipping Weight (Metric)45.9 kg
Electrical Requirements115 V 50/60 Hz
Voltage115 V
Amperage12.6 A