▸
Tubes
▸
Tube Caps and Closures
▸
Flanged Plug Caps For 12mm Tubes
Actual product may vary
Globe Scientific Inc
Flanged Plug Caps For 12mm Tubes
Provide leak-resistance in test tubes
Have Questions?
Change view
Catalog Number
Color
FIS22-171-666
Orange
FIS22-171-664
Black
FIS22-171-665
Lavender
3 Options
Catalog number
FIS22-171-666
Price (SGD)
60.00
Pack
Add to cart
Color:
Orange
Price (SGD)
60.00
Pack
Add to cart
Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
Available in nine colors for sample identification purposes
Specifications
Includes
2 flexible flanges
Product Type
Flange Plug Cap
Material
Polyethylene
Color
Orange
Size (English)
0.47 in.
Size (Metric)
12 mm
For Use With (Equipment)
12 mm Tubes