Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Tubes
▸
Test Tubes
▸
Disposable Test Tubes
▸
16 x 100mm Test Tubes
Actual product may vary
Globe Scientific Inc
16 x 100mm Test Tubes
Molded from premium-grade polystyrene for clarity
Have Questions?
Catalog Number
Graduated
Packaging
Rimmed
Material
FIS22-171-621
No
2000/Cs.
No
Polystyrene
Catalog number
FIS22-171-621
Price (SGD)
451.00
Case
Add to cart
Graduated:
No
Packaging:
2000/Cs.
Rimmed:
No
Material:
Polystyrene
Price (SGD)
451.00
Case
Add to cart
Molded from premium-grade polystyrene for clarity
Used for laboratory applications
Volume: 12mL
Max. Force: 1500 RPM
Freezable
Polystyrene tubes
Withstand 1500rpm during centrifugation
Specifications
Capacity (Metric)
12 mL
Length (Metric)
100 mm
Material
Polystyrene
Shape
Round Bottom
Packaging
2000/Cs.
Rimmed
No
Autoclavable
Not Autoclavable
Graduated
No
Diameter (Metric)
16 mm
Product Type
Test Tube