17 x 100mm Test Tubes
Actual product may vary
Globe Scientific Inc
17 x 100mm Test Tubes
Molded from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis and solvents
Have Questions?
Catalog NumberMaterialGraduated
FIS22-171-615PolypropyleneNo
Catalog number FIS22-171-615
Price (SGD)
267.00
Case
Material:
Polypropylene
Graduated:
No
Price (SGD)
267.00
Case
  • Available in Polypropylene and Polystyrene
  • Used for laboratory applications
  • Come in wide range of sizes
Polystyrene tubes
Withstand 1500rpm during centrifugation
Polypropylene tubes
Withstand 3000rpm during centrifugation
Autoclavable and freezable
Specifications
Capacity (Metric)14 mL
Length (Metric)100 mm
MaterialPolypropylene
SterilityNon-sterile
AutoclavableAutoclavable
GraduatedNo
Diameter (Metric)17 mm
Product TypeTest Tube
Unit SizeCase