Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Pipet Products
▸
Transfer Pipets
▸
Graduated Transfer Pipets
Actual product may vary
Globe Scientific Inc
Graduated Transfer Pipets
Comes with 7mL, large bulb
Have Questions?
Catalog Number
Quantity
FIS22-170-404
5000/Cs.
Catalog number
FIS22-170-404
Price (SGD)
49.00
Pack
Add to cart
Quantity:
5000/Cs.
Price (SGD)
49.00
Pack
Add to cart
Made from LDPE
Graduated up to 3mL
6.3 in. (160mm) long
Chemically resistant to dilute and concentrated acids, alcohols, bases and esters
Non-sterile and non-autoclavable
Specifications
Bulb Draw (Metric)
3.2 mL
Diameter (Metric) Outer
0.79 cm
Material
LDPE
Sterility
Non-sterile
Drops/mL
20
Graduated
Yes
Graduations
3 mL
Quantity
5000/Cs.
Volume (Metric)
7 mL
Length (Metric)
16 cm
Type
Transfer Pipet