Polypropylene 15 mL Centrifuge Tubes
Actual product may vary
Globe Scientific Inc
Polypropylene 15 mL Centrifuge Tubes
Feature flat-top, plug-seal leak resistant screw caps, large white writing area and printed graduation marks
Have Questions?
Catalog NumberSterilityNo. per CasePackaging
FIS22-170-256Non-sterile500Bulk
Catalog number FIS22-170-256
Price (SGD)
455.00
Case
Sterility:
Non-sterile
No. per Case:
500
Packaging:
Bulk
Price (SGD)
455.00
Case
  • Spin Rate: 10,000 rcf (12,320 x g)
  • Material: Polypropylene (PP)
  • Cap is made from High Density Polyethylene (HDPE)
  • Dimensions: 17 x 120 mm
  • White graduation marks and large white writing area
Specifications
Closure ColorRed
Capacity (Metric)15 mL
MaterialPolypropylene
No. per Case500
PackagingBulk
SterilityNon-sterile
Length (Metric)120 mm
Bottom ShapeConical
Closure TypeScrew Cap
Closure IncludedYes
Diameter (Metric)17 mm
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF12,320 x g
Unit SizeCase