Flanged Plug Tube Caps
Actual product may vary
Globe Scientific Inc
Flanged Plug Tube Caps
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
Have Questions?
Catalog NumberColorSize (Metric)
FIS22-170-148Clear16 mm
Catalog number FIS22-170-148
Price (SGD)
68.00
Pack
Color:
Clear
Size (Metric):
16 mm
Price (SGD)
68.00
Pack
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
These flanged plug caps are designed for test tubes, centrifuge tubes, and round cuvettes, providing a secure seal through a dual-flange design that offers superior leak resistance compared to standard plug caps.
  • Cap Style: Flanged plug cap
  • Size: 16 mm
  • Material: Polyethylene (PE)
  • Color: Clear
Specifications
Product TypeFlange Plug Cap
MaterialPolyethylene
ColorClear
Size (English)0.62 in.
Size (Metric)16 mm
For Use With (Equipment)16 mm Tubes
Unit SizePack