Water Sample Dipper
Actual product may vary
Bel-Art
Water Sample Dipper
Long reach for otherwise inaccessible spots or taking samples of water, sewage, wastes and effluents from lakes, streams, tanks or ponds
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-1638571000 mL
Catalog number FIS22-163857
Price (SGD)
700.00
Each
Capacity (Metric):
1000 mL
Price (SGD)
700.00
Each
Long reach for otherwise inaccessible spots or taking samples of water, sewage, wastes and effluents from lakes, streams, tanks or ponds
  • The handle has 3 parts, a 91 cm (3 ft.) lower section, a 112 cm (44 in.) telescoping section, and an additional 91 cm (3 ft.) upper section for an extended reach
  • Connections are made with spring buttons with adjusting steps 15 cm (6 in.) apart
  • Minimum handle length is 91 cm (3 ft.), maximum length is 274 cm (108 in.)
  • Handle diameter is 2.5 cm (1 in.) with 2.2 cm (0.875 in.) telescoping tube
  • A 1000 mL graduated polypropylene pitcher is mounted on an aluminum tube handle
Specifications
Product TypeDip Sampler
MaterialPolypropylene
AutoclavableNot Autoclavable
Capacity (English)33.81 oz.
Capacity (Metric)1000 mL
For Use With (Application)Safely reach water samples in tanks, lakes, ponds
Length (English)3 ft.
Length (Metric)0.9 m
Unit SizeEach