▸
Hotplates and Stirrers
▸
Stirrer Accessories
▸
Stirring Paddles
Actual product may vary
Bel-Art
Stirring Paddles
For hand or automated mixing liquids in large vats and tanks
Have Questions?
Catalog Number
Length (Metric) Handle
FIS22-163816
91 cm
Catalog number
FIS22-163816
Price (SGD)
330.00
Each
Add to cart
Length (Metric) Handle:
91 cm
Price (SGD)
330.00
Each
Add to cart
Made from high density polyethylene
Built-in hang up hook to store tool
Specifications
For Use With (Application)
Hand or automated mixing liquids in large vats and tanks
Length (English) Handle
36 in.
Length (Metric) Handle
91 cm
Type
Paddle Stirrer
Material
Polyethylene