Pipetter Nozzle Filter Kits
Actual product may vary
VistaLab Technologies
Pipetter Nozzle Filter Kits
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
Have Questions?
Catalog NumberVolume (Metric)
FIS22-146-68410 to 100 μL
Catalog number FIS22-146-684
Price (SGD)
152.00
Pack
Volume (Metric):
10 to 100 μL
Price (SGD)
152.00
Pack
  • Available for small or large tips
Specifications
DescriptionLarge Nozzle
For Use WithPipettes
MaterialPlastic
SterilityNon-sterile
TypeNozzle Replacement Filter Kit
Volume (Metric)10 to 100 μL
For Use With (Equipment)25 to 1250 μL Electronic Single Channel Model, 100 to 1000 μL Adjustable Model, 201 to 1000 μL, 25 Filters Fixed Models
Quantity1 Ea.
Length (English)5 in.
Width (English)3 in.
Height (English)1 in.
Unit SizePack