Reagent Reservoirs
Actual product may vary
VistaLab Technologies
Reagent Reservoirs
Unique trough-within-trough design maximizes reagent recovery
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityPackagingQuantity
FIS22-145-10125 mLNon-sterileBulk10/bag x 10; 100/Cs.
FIS21-381-27A100 mLSterileBulk10/bag x 10; 100/Cs.
FIS21-381-27E25 mL (divided)SterileBulk10/bag x 10; 100/Cs.
FIS21-381-27D25 mLSterileIndividually Wrapped50/Cs.
FIS21-381-27G50 mLSterileBulk10/bag x 10; 100/Cs.
FIS2138127G50 mLSterileBulk10/bag x 10; 100/Cs.
FIS2138127H50 mLSterileIndividually Wrapped50/Cs.
FIS21-385-101100 mLSterileIndividually Wrapped100/Cs.
FIS21-385-10225 mLSterileBulk20/bag x 10; 200/Cs.
FIS21-385-10550 mLSterileIndividually Wrapped100/Cs.
FIS21-385-10325 mLSterileIndividually Wrapped100/Cs.
FIS22-145-10350 mLNon-sterileBulk10/bag x 10; 100/Cs.
FIS22-145-100100 mLNon-sterileBulk10/bag x 10; 100/Cs.
FIS21-381-27C25 mLSterileBulk10/bag x 10; 100/Cs.
FIS21-381-09010 mLSterileBulk100/Cs.
Catalog number FIS22-145-101
Price (SGD)
281.00
Case
Capacity (Metric):
25 mL
Sterility:
Non-sterile
Packaging:
Bulk
Quantity:
10/bag x 10; 100/Cs.
Price (SGD)
281.00
Case
  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Specifications
Certifications/CompliancePyrogen-free
ColorWhite
Capacity (Metric)25 mL
MaterialPolystyrene
SterilityNon-sterile
TypeReagent Reservoir
For Use With (Application)General Liquid Handling
Height (English)7.5 in.
Length (English)13 in.
PackagingBulk
Quantity10/bag x 10; 100/Cs.
Width (English)7 in.
AutoclavableNot Autoclavable
Channels1
DisposableSingle-use
Bottom ShapeV-bottom
Unit SizeCase