Aqui-Pad™ Benchtop Absorbent Mat
Actual product may vary
Therapak
Aqui-Pad™ Benchtop Absorbent Mat
Designed for use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work.
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-131-401750 mL
Catalog number FIS22-131-401
Price (SGD)
323.00
Case
Capacity (Metric):
750 mL
Price (SGD)
323.00
Case
  • Consists of two material layers with absorbent media sealed inside
  • Non-woven polypropylene pad top layer and impervious polypropylene film bottom layer to enclose any liquid spills so the liquid does not reach the benchtop
  • Inside the two layers is a proprietary absorbent media fabricated from tissue paper and wood pulp materials that are impregnated with a super absorbent polymer
  • Supplied in flat sheets 18 x 11 in. (457 x 280mm)
  • Absorbs up to 750mL
  • Placed in biohazard disposal after use
Specifications
Capacity (English)0.19 gal.
Capacity (Metric)750 mL
MaterialPolypropylene
TypeAqui-Pad™ Benchtop Absorbent Mat
For Use With (Application)For use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work
Unit SizeCase