ConstanTemp™ Specimen Insulator for Lock Boxes
Actual product may vary
Therapak
ConstanTemp™ Specimen Insulator for Lock Boxes
An ideal source of protection for maintaining specimen integrity prior to courier pick up in lock boxes or in courier transit
Have Questions?
Catalog NumberColor
FIS22-130-218Blue
Catalog number FIS22-130-218
Price (SGD)
920.00
Case
Color:
Blue
Price (SGD)
920.00
Case
Features a fabric-based construction and Volara insulation combined with specially formulated gel that holds temperature for extended periods of time. For frozen specimens, place ConstanTemp in the freezer overnight. When specimen is frozen, insert it into the pouch and place in the lock box for transport. It will stay frozen over a period of 19 hours at an ambient temperature of 22°C. For refrigerated specimens, place insulator into refrigerator overnight or until it reaches 2°C. It will maintain 2–8°C in either a hot or cold environment for hours. For room temperature or ambient specimens, leave insulator on the counter in a 20-25°C room and place specimen inside. It will maintain this temperature for hours.
  • Maintains ambient, refrigerated and frozen specimen integrity
  • Custom printing and fabric color options available
  • Blue color (also available in red and white; contact your Fisher HealthCare representative)
Specifications
ColorBlue
Unit SizeCase