Zip Bag with Biohazard Symbol
Actual product may vary
Zip Bag with Biohazard Symbol
Zip Bag with Biohazard Symbol
Therapak
Zip Bag with Biohazard Symbol
For large capacity shipments of multiple sample packages from a single site or multiple sites to a testing laboratory
Have Questions?
Catalog NumberDimensions (L x W)
FIS22-130-11113 x 18 in.
Catalog number FIS22-130-111
Price (SGD)
597.00
Case
Dimensions (L x W):
13 x 18 in.
Price (SGD)
597.00
Case
Specifications
AutoclavableNot Autoclavable
ColorClear
For Use With (Application)Used for large capacity shipments of multiple sample packages from a single site or multiple sites to a testing laboratory
IncludesBiohazard symbol, zip top, 2 pockets
MaterialLDPE
Dimensions (L x W)13 x 18 in.
Product TypeBiohazard Transport Bag
Length (Metric)328 mm
Width (Metric)485 mm
Unit SizeCase