Dry Ice Label
Actual product may vary
Therapak
Dry Ice Label
Complies with DOT and IATA regulations for all dry ice shipments
Have Questions?
Catalog NumberSize (Metric)
FIS22-130-06513.9 x 13.9 cm
Catalog number FIS22-130-065
Price (SGD)
257.00
Pack
Size (Metric):
13.9 x 13.9 cm
Price (SGD)
257.00
Pack
  • Self-adhesive roll stock for peel and apply convenience
  • Preprinted with all required information—simply complete designated Shipper and Consignee name and address, amount of dry ice (in kg) included in package, and apply label to package
Specifications
Certifications/ComplianceDOT 49 CFR 173.217, IATA
Adhesive TypeSelf-adhesive
FormatLabel
Product TypeDry Ice Shipping Label
For Use With (Application)Used to transport biological specimens
Quantity500/Pk.
Size (English)5.5 x 5.5 in.
Size (Metric)13.9 x 13.9 cm
Length (English)5.5 in.
Width (English)5.5 in.
Unit SizePack