Absorbent Materials
Actual product may vary
Therapak
Absorbent Materials
For proper packing of biological specimens in accordance with IATA and DOT regulations
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeSizeCapacity (Metric)
FIS22-130-044Absorbent Wadding12 in. x 100 ft.Absorbent: 400 mL
FIS22-130-040Absorbent Sheet3 x 6 in.Absorbent: 120 mL
FIS22-130-041Absorbent Sheet6 x 6 in.Absorbent: 240 mL
Catalog number FIS22-130-044
Price (SGD)
581.00
Each
Type:
Absorbent Wadding
Size:
12 in. x 100 ft.
Capacity (Metric):
Absorbent: 400 mL
Price (SGD)
581.00
Each
For proper packing of biological specimens in accordance with IATA and DOT regulations
Absorbent Sheets
  • Tissue-based material impregnated with super-absorbent polymers
  • Place between primary receptacle and secondary packaging
  • Available in 3 x 4 in., 3 x 6 in., 6 x 6 in. or 12 x 12 in.
Absorbent Wadding
  • 12 in. perforated sections on a roll
  • Excellent void fill and cushioning material for biological specimen transport
  • Protects fragile shipping containers and specimen tubes
  • Each 12 x 12 in. section is capable of absorbing up to 400 mL of liquid
To comply with IATA Packing Instructions 650and DOT Triple Packaging Requirements, insert sufficient absorbent material between primary receptacle and secondary packaging to contain the total potential volume of the specimen(s) being transported.
Specifications
ApplicationsFor Biological specimen transport
Size12 in. x 100 ft.
TypeAbsorbent Wadding
Capacity (Metric)Absorbent: 400 mL
Unit SizeEach