Product Image
Therapak
Bag Specimen 7 W x 11 L in. 100 pcs
Bag Specimen 7 W x 11 L in. 100 pcs
Have Questions?
Catalog number FIS22-130-021
Price (SGD)
474.00
Case
Price (SGD)
474.00
Case
Bag Specimen 7 W x 11 L in. 100 pcs
Specifications
Certifications/ComplianceMeet DOT and IATA regulations for shipping biological substances.
ColorClear with White Tape Liner
For Use With (Application)For Use as a Secondary Water-Tight Receptacle
MaterialPlastic
Dimensions (L x W)7 x 11 in.
Length (English)11 in.
Width (English)7 in.
Product TypeCompliant Transport Bags
Length (Metric)27.9 cm
Width (Metric)17.8 cm
Unit SizeCase